5-Bromo-2-fluoronicotinaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-fluoropyridine-3-carboxaldehyde
PC11103-1G PC11103-5G
APOSPC11103-1GEA 21.9 EUR
APOSPC11103-1G APOSPC11103-5G
5-Bromo-2-fluoronicotinaldehyde
5-Bromo-2-fluoronicotinaldehyde
Formula: C₆H₃BrFNO
MW: 204.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09842440
CAS Number: 875781-15-0