3,5-Difluorosalicylaldehyde 98%
Supplier: Apollo Scientific
Warning
PC1112-1G PC1112-5G PC1112-25G
APOSPC1112-1GEA 36.4 EUR
APOSPC1112-1G APOSPC1112-5G APOSPC1112-25G
3,5-Difluorosalicylaldehyde 98%
3,5-Difluorosalicylaldehyde
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD04039224
CAS Number: 63954-77-8