4'-Fluorobiphenyl-4-carboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(4-Fluorophenyl)benzaldehyde
PC11137-1G
PC11137-250MG
PC11137-5G
APOSPC11137-1GEA
94.5
EUR
APOSPC11137-1G
APOSPC11137-250MG
APOSPC11137-5G
4'-Fluorobiphenyl-4-carboxaldehyde
4'-Fluorobiphenyl-4-carboxaldehyde
Formula:
C₁₃H₉FO MW: 200.212 g/mol Boiling Pt: 324 °C Melting Pt: 93…97 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631912 CAS Number: 60992-98-5 |