4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde
PC11174-1G PC11174-250MG
APOSPC11174-1GEA 31.9 EUR
APOSPC11174-1G APOSPC11174-250MG
4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde
4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde
Formula: C₁₄H₉F₃O
MW: 250.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631879
CAS Number: 343604-24-0