3'-(Trifluoromethyl)biphenyl-3-carboxylic acid 96%
Supplier: Apollo Scientific
Warning
PC11182-1G PC11182-5G
APOSPC11182-1GEA 266 EUR
APOSPC11182-1G APOSPC11182-5G
3'-(Trifluoromethyl)biphenyl-3-carboxylic acid 96%
3'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424603
CAS Number: 168619-05-4