[2-Fluoro-4-(methylsulphonyl)phenyl]hydrazine 95+%
Supplier: Apollo Scientific
Warning
PC11202-1G PC11202-5G
APOSPC11202-1GEA 81.2 EUR
APOSPC11202-1G APOSPC11202-5G
[2-Fluoro-4-(methylsulphonyl)phenyl]hydrazine 95+%
[2-Fluoro-4-(methylsulphonyl)phenyl]hydrazine
Formula: C₇H₉FN₂O₂S
MW: 204.22 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037953
CAS Number: 832714-48-4