2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethanone 95+%
Supplier: Apollo Scientific
Warning
PC11204-1G PC11204-5G PC11204-25G
APOSPC11204-1GEA 163 EUR
APOSPC11204-1G APOSPC11204-5G APOSPC11204-25G
2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethanone 95+%
2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethanone
Formula: C₁₆H₁₀F₃NO₂
MW: 305.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037916
CAS Number: 849021-37-0