1,2-Difluoro-4-(methylsulphonyl)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Difluorophenyl methyll sulphone
PC11206-10G PC11206-1G PC11206-5G
APOSPC11206-10GEA 34.8 EUR
APOSPC11206-10G APOSPC11206-1G APOSPC11206-5G
1,2-Difluoro-4-(methylsulphonyl)benzene
1,2-Difluoro-4-(methylsulphonyl)benzene
Formula: C₇H₆F₂O₂S
MW: 192.1800 g/mol
Melting Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD04037934
CAS Number: 424792-57-4