3-Bromo-5-fluorobenzonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
PC1121-25G
PC1121-5G
APOSPC1121-25GEA
66.7
EUR
APOSPC1121-25G
APOSPC1121-5G
3-Bromo-5-fluorobenzonitrile 98%
3-Bromo-5-fluorobenzonitrile
Formula:
C₇H₃BrFN Melting Pt: 43…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038227 CAS Number: 179898-34-1 UN: 3439 ADR: 6.1,III |