1,3-Dichloro-2-fluoro-5-nitrobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dichloro-4-fluoro-1-nitrobenzene
PC11210-25G PC11210-100G PC11210-500G
APOSPC11210-25GEA 21.9 EUR
APOSPC11210-25G APOSPC11210-100G APOSPC11210-500G
1,3-Dichloro-2-fluoro-5-nitrobenzene 99%
1,3-Dichloro-2-fluoro-5-nitrobenzene
Formula: C₆H₂Cl₂FNO₂
MW: 209.99 g/mol
Melting Pt: 44 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00129963
CAS Number: 3107-19-5