Potassium-3-fluoro-4-phenylphenyltrifluoroborate
Supplier: Apollo Scientific
Warning
Synonyms: Potassium (2-fluorobiphenyl-4-yl)trifluoroborate, Potassium trifluoro(2-fluorobiphenyl-4-yl)borate
PC11234-1G PC11234-5G
APOSPC11234-1GEA 113 EUR
APOSPC11234-1G APOSPC11234-5G
Potassium-3-fluoro-4-phenylphenyltrifluoroborate
Potassium 3-fluoro-4-phenylphenyltrifluoroborate
Formula: C₁₂H₈BF₄K
Melting Pt: 290…294 °C
Storage Temperature: Ambient
MDL Number: MFCD08235035
CAS Number: 850623-57-3