4,5-Difluoro-2-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Danger
PC1124-10G PC1124-5G PC1124-1G
APOSPC1124-10GEA 215 EUR
APOSPC1124-10G APOSPC1124-5G APOSPC1124-1G
4,5-Difluoro-2-methoxybenzonitrile 98%
4,5-Difluoro-2-methoxybenzonitrile
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD06660346
CAS Number: 425702-28-9
UN: 3439
ADR: 6.1,III