2-Chloro-N-(3,4-difluorophenyl)acetamide
Supplier: Apollo Scientific
Warning
Synonyms: N-Chloroacetyl-3,4-difluoroaniline
PC11253-1G PC11253-250MG
APOSPC11253-1GEA 55.3 EUR
APOSPC11253-1G APOSPC11253-250MG
2-Chloro-N-(3,4-difluorophenyl)acetamide
2-Chloro-N-(3,4-difluorophenyl)acetamide
Formula: C₈H₆ClF₂NO
MW: 205.59 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01354030
CAS Number: 76778-13-7