4-Bromo-α,α,α,2-tetrafluorotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-fluorobenzotrifluoride, 1-Bromo-3-fluoro-4-(trifluoromethyl)benzene
PC11257-25G PC11257-5G
APOSPC11257-25GEA 49.3 EUR
APOSPC11257-25G APOSPC11257-5G
4-Bromo-α,α,α,2-tetrafluorotoluene
4-Bromo-α,α,α,2-tetrafluorotoluene
Formula: C₇H₃BrF₄
Density: 1.72 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD01075254
CAS Number: 142808-15-9