Pentafluorophenyl-4-bromobenzenesulphonate
Supplier: Apollo Scientific
PC11274-1G PC11274-500MG PC11274-5G
APOSPC11274-1GEA 275 EUR
APOSPC11274-1G APOSPC11274-500MG APOSPC11274-5G
Pentafluorophenyl-4-bromobenzenesulphonate
Pentafluorophenyl 4-bromobenzenesulphonate
Formula: C₁₂H₄BrF₅O₃S
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00404028
CAS Number: 848649-38-7