3,5-Difluoro-2-methoxybenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC1129-10G
PC1129-5G
PC1129-1G
APOSPC1129-10GEA
191
EUR
APOSPC1129-10G
APOSPC1129-5G
APOSPC1129-1G
3,5-Difluoro-2-methoxybenzaldehyde 98%
3,5-Difluoro-2-methoxybenzaldehyde
Formula:
C₈H₆F₂O₂ Melting Pt: 39…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD06246883 CAS Number: 131782-50-8 |