2,6-Difluoro-3-hydroxybenzaldehyde
Supplier: Apollo Scientific
Warning
PC1132-1G PC1132-500MG PC1132-5G
APOSPC1132-1GEA 58 EUR
APOSPC1132-1G APOSPC1132-500MG APOSPC1132-5G
2,6-Difluoro-3-hydroxybenzaldehyde
2,6-Difluoro-3-hydroxybenzaldehyde
Formula: C₇H₄F₂O₂
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD08275284
CAS Number: 152434-88-3