3,5-Difluoro-2-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Danger
PC1136-1G PC1136-5G PC1136-25G PC1136-100G
APOSPC1136-1GEA 26.3 EUR
APOSPC1136-1G APOSPC1136-5G APOSPC1136-25G APOSPC1136-100G
3,5-Difluoro-2-methoxybenzonitrile 98%
3,5-Difluoro-2-methoxybenzonitrile
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD06409085
CAS Number: 874804-08-7
UN: 3439
ADR: 6.1 III