3,6-Difluoro-2-methoxybenzaldehyde 98%
Supplier: Apollo Scientific
PC1143-1G PC1143-5G PC1143-25G
APOSPC1143-1GEA 30.6 EUR
APOSPC1143-1G APOSPC1143-5G APOSPC1143-25G
3,6-Difluoro-2-methoxybenzaldehyde 98%
3,6-Difluoro-2-methoxybenzaldehyde
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275289
CAS Number: 887267-04-1