2-Bromo-3'-fluoro-4'-methoxyacetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluoro-4-methoxyphenacyl bromide
PC1155-10G PC1155-1G PC1155-5G
APOSPC1155-10GEA 719 EUR
APOSPC1155-10G APOSPC1155-1G APOSPC1155-5G
2-Bromo-3'-fluoro-4'-methoxyacetophenone
2-Bromo-3'-fluoro-4'-methoxyacetophenone
Formula: C₉H₈BrFO₂
Melting Pt: 71…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00466240
CAS Number: 350-27-6
UN: 1759
ADR: 8,III