4-Ethynyl-1,2-difluorobenzene 95%
Supplier: Apollo Scientific
Danger
Synonyms: 3,4-Difluorophenylacetylene
PC1156-5G PC1156-1G PC1156-25G
APOSPC1156-5GEA 200 EUR
APOSPC1156-5G APOSPC1156-1G APOSPC1156-25G
4-Ethynyl-1,2-difluorobenzene 95%
4-Ethynyl-1,2-difluorobenzene
Formula: C₈H₄F₂
MW: 138.12 g/mol
Boiling Pt: 54 °C/2mmHg
Density: 1.144 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD08059511
CAS Number: 143874-13-9
UN: 1993
ADR: 3,II