2-Chloro-6-trifluoromethyl-3-(trimethylsilyl)pyridine
Supplier: Apollo Scientific
PC1182-1G PC1182-5G
APOSPC1182-1GEA 199 EUR
APOSPC1182-1G APOSPC1182-5G
2-Chloro-6-trifluoromethyl-3-(trimethylsilyl)pyridine
2-Chloro-6-trifluoromethyl-3-(trimethylsilyl)pyridine
Formula: C₉H₁₁ClF₃NSi
MW: 253.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09763705
CAS Number: 205444-18-4