4-Fluoro-3-nitrobenzonitrile
Supplier: Apollo Scientific
Warning
PC1197-100G PC1197-25G PC1197-5G
APOSPC1197-100GEA 174 EUR
APOSPC1197-100G APOSPC1197-25G APOSPC1197-5G
4-Fluoro-3-nitrobenzonitrile
4-Fluoro-3-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01632197
CAS Number: 1009-35-4
UN: 3439
ADR: 6.1 III