4,4,4-Trifluoro-1-phenyl-1,3-butanedione 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Benzoyl-1,1,1-trifluoroacetone, (4,4,4-Trifluor-1,3-butandionyl)benzol, 4,4,4-Trifluoro-1-phenylbutane-1,3-dione, Benzoyl-1,1,1-trifluoroacetone
PC1210-1KG PC1210-500G PC1210-100G
APOSPC1210-1KGEA 836 EUR
APOSPC1210-1KG APOSPC1210-500G APOSPC1210-100G
4,4,4-Trifluoro-1-phenyl-1,3-butanedione 97%
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Boiling Pt: 224 °C
Melting Pt: 37…40 °C
Density: 1.113 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000425
CAS Number: 326-06-7
EINECS: 206-307-8