3-Fluoropyridine-2-carbonitrile 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluoro-2-pyridinecarbonitrile, 2-Cyano-3-fluoropyridine, 3-Fluoropicolinonitrile
PC1215-5G PC1215-25G PC1215-100G
APOSPC1215-5GEA 23.2 EUR
APOSPC1215-5G APOSPC1215-25G APOSPC1215-100G
3-Fluoropyridine-2-carbonitrile 97%
3-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
Boiling Pt: 120…125 °C/17mmHg
Melting Pt: 27…32 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD06797501
CAS Number: 97509-75-6
UN: 3439
ADR: 6.1 III