4-(Trifluoromethyl)salicylic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-4-(trifluoromethyl)benzoic acid
PC1216-100G PC1216-25G PC1216-500G
APOSPC1216-100GEA 106 EUR
APOSPC1216-100G APOSPC1216-25G APOSPC1216-500G
4-(Trifluoromethyl)salicylic acid 98%
4-(Trifluoromethyl)salicylic acid
Formula: C₈H₅F₃O₃
MW: 206.1187 g/mol
Melting Pt: 178…179 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD01529657
CAS Number: 328-90-5