2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane 99%
Supplier: Apollo Scientific
Warning
Synonyms: 5,5'-(Hexafluoroisopropylidene)di-o-toluidine
PC1224H-25G PC1224H-50G PC1224H-5G
APOSPC1224H-25GEA 328 EUR
APOSPC1224H-25G APOSPC1224H-50G APOSPC1224H-5G
2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane 99%
2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane
Formula: C₁₇H₁₆F₆N₂
MW: 362.32 g/mol
Melting Pt: 105…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00039148
CAS Number: 116325-74-7