2,2-Bis(4-carboxyphenyl)hexafluoropropane 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4'-(Hexafluoroisopropylidene)bis(benzoic Acid)
PC1226-100G
APOSPC1226-100GEA 831 EUR
APOSPC1226-100G
2,2-Bis(4-carboxyphenyl)hexafluoropropane 98%
2,2-Bis(4-carboxyphenyl)hexafluoropropane
Formula: C₁₇H₁₀F₆O₄
MW: 392.25 g/mol
Melting Pt: 272…275 °C
Density: 1.504 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040931
CAS Number: 1171-47-7