4,4'-(Hexafluoroisopropylidene)diphenol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol
PC1229-100G PC1229-250G PC1229-1KG
APOSPC1229-100GEA 37.7 EUR
APOSPC1229-100G APOSPC1229-250G APOSPC1229-1KG
4,4'-(Hexafluoroisopropylidene)diphenol 98%
4,4'-(Hexafluoroisopropylidene)diphenol
Formula: C₁₅H₁₀F₆O₂
MW: 336.24 g/mol
Boiling Pt: 350…400 °C
Melting Pt: 160…163 °C
Density: 1.53 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000439
CAS Number: 1478-61-1
EINECS: 216-036-7
UN: 3077
ADR: 9+EHS,III