Bis(perfluoroisopropyl)ketone 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Bis(perfluoroisopropyl)methanone
PC1242-1G
PC1242-5G
PC1242-250MG
APOSPC1242-1GEA
174
EUR
APOSPC1242-1G
APOSPC1242-5G
APOSPC1242-250MG
Bis(perfluoroisopropyl)ketone 97%
Bis(perfluoroisopropyl)ketone
Formula:
C₇F₁₄O MW: 366.06 g/mol Boiling Pt: 72…73 °C Density: 1.661 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042087 CAS Number: 813-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...