3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid 95%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Hydroxy-2,2-bis(trifluoromethyl)acetic Acid
PC1253T-1G PC1253T-5G PC1253T-25G
APOSPC1253T-1GEA 140 EUR
APOSPC1253T-1G APOSPC1253T-5G APOSPC1253T-25G
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid 95%
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid
Formula: C₄H₂F₆O₃
MW: 212.05 g/mol
Boiling Pt: 153…156 °C
Melting Pt: 77…84 °C
Density: 1.183 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00190631
CAS Number: 662-22-6
UN: 3261
ADR: 8 III