3,4-Dibromo-1,1,1,2,2-pentafluorobutane 97%
Supplier: Apollo Scientific
Warning
PC1269-1G PC1269-5G
APOSPC1269-1GEA 105 EUR
APOSPC1269-1G APOSPC1269-5G
3,4-Dibromo-1,1,1,2,2-pentafluorobutane 97%
3,4-Dibromo-1,1,1,2,2-pentafluorobutane
Formula: C₄H₃Br₂F₅
Melting Pt: 123 °C
Density: 1.994 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094110
CAS Number: 378-69-8