1-Chloro-2-fluoro-4-nitrobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-fluoronitrobenzene
PC1278-5G PC1278-100G PC1278-25G
APOSPC1278-5GEA 21.9 EUR
APOSPC1278-5G APOSPC1278-100G APOSPC1278-25G
1-Chloro-2-fluoro-4-nitrobenzene
1-Chloro-2-fluoro-4-nitrobenzene
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Boiling Pt: 114…116 °C/24mmHg
Melting Pt: 63…64 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00215833
CAS Number: 350-31-2