2,2,3,3-Tetrafluorocyclobutanol
Supplier: Apollo Scientific
Warning
PC1328-1G PC1328-5G
APOSPC1328-1GEA 264 EUR
APOSPC1328-1G APOSPC1328-5G
2,2,3,3-Tetrafluorocyclobutanol
2,2,3,3-Tetrafluorocyclobutanol
Formula: C₄H₄F₄O
MW: 144.07 g/mol
Boiling Pt: 152…157 °C
Density: 1.414 g/cm³ (25 °C)
Flash Pt: 44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190085
CAS Number: 374-32-3
UN: 1987
ADR: 3,III