4-Fluororesorcinol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-2,4-dihydroxybenzene
PC1341-10G PC1341-25G PC1341-5G
APOSPC1341-10GEA 288 EUR
APOSPC1341-10G APOSPC1341-25G APOSPC1341-5G
4-Fluororesorcinol 98%
4-Fluororesorcinol
Formula: C₆H₅FO₂
MW: 128.10 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Freezer
MDL Number: MFCD03789074
CAS Number: 103068-41-3