6-Amino-2-fluoro-9H-purine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-9H-purin-6-amine, 2-Fluoroadenine
PC1349-1G PC1349-250MG PC1349-5G
APOSPC1349-1GEA 29 EUR
APOSPC1349-1G APOSPC1349-250MG APOSPC1349-5G
6-Amino-2-fluoro-9H-purine
6-Amino-2-fluoro-9H-purine
Formula: C₅H₄FN₅
MW: 153.12 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD01940409
CAS Number: 700-49-2