4-Bromo-α,α,α-trifluorotoluene 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene
PC1350-100G
PC1350-1KG
PC1350-500G
APOSPC1350-100GEA
56.7
EUR
APOSPC1350-100G
APOSPC1350-1KG
APOSPC1350-500G
4-Bromo-α,α,α-trifluorotoluene 99%
4-Bromo-α,α,α-trifluorotoluene
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol Boiling Pt: 154…156 °C Density: 1.607 g/cm³ (25 °C) Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000398 CAS Number: 402-43-7 EINECS: 206-943-6 UN: 1993 ADR: 3 III |