4-Bromo-3-fluoroanisole 98%
Supplier: Apollo Scientific
Warning
PC1357-25G PC1357-5G PC1357-1G
APOSPC1357-25GEA 49.3 EUR
APOSPC1357-25G APOSPC1357-5G APOSPC1357-1G
4-Bromo-3-fluoroanisole 98%
4-Bromo-3-fluoroanisole
Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 216 °C
Density: 1.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01310983
CAS Number: 458-50-4