5-Bromo-2-chlorobenzotrifluoride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-chloro-3-(trifluoromethyl)benzene, 5-Bromo-2-chlorobenzotrifluoride, 4-Bromo-1-chloro-2-(trifluoromethyl)benzene, 5-Bromo-2-chloro-α,α,α-trifluorotoluene
PC1359-100G PC1359-25G
APOSPC1359-100GEA 29 EUR
APOSPC1359-100G APOSPC1359-25G
5-Bromo-2-chlorobenzotrifluoride 98%
5-Bromo-2-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 195 °C
Melting Pt: –21 °C
Density: 1.745 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000601
CAS Number: 445-01-2
EINECS: 207-149-2