2-Bromo-3-fluorotoluene 98%
Supplier: Apollo Scientific
Warning
PC1362-100G PC1362-25G PC1362-5G
APOSPC1362-100GEA 213 EUR
APOSPC1362-100G APOSPC1362-25G APOSPC1362-5G
2-Bromo-3-fluorotoluene 98%
2-Bromo-3-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 187 °C
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD08458010
CAS Number: 59907-13-0