2-Bromo-4-chloro-6-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC1365-25G PC1365-5G PC1365-100G
APOSPC1365-25GEA 143 EUR
APOSPC1365-25G APOSPC1365-5G APOSPC1365-100G
2-Bromo-4-chloro-6-fluoroaniline 98%
2-Bromo-4-chloro-6-fluoroaniline
Formula: C₆H₄BrClFN
Melting Pt: 41…43 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00070746
CAS Number: 195191-47-0