2-Bromo-6-chloro-4-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC1366-25G PC1366-5G
APOSPC1366-25GEA 75.4 EUR
APOSPC1366-25G APOSPC1366-5G
2-Bromo-6-chloro-4-fluoroaniline 98%
2-Bromo-6-chloro-4-fluoroaniline
Formula: C₆H₄BrClFN
Melting Pt: 54…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051707
CAS Number: 201849-14-1