1-Bromo-3-chloro-4-fluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-chloro-1-fluorobenzene, 4-Bromo-2-chlorofluorobenzene
PC1367-250G PC1367-100G PC1367-500G
APOSPC1367-250GEA 109 EUR
APOSPC1367-250G APOSPC1367-100G APOSPC1367-500G
1-Bromo-3-chloro-4-fluorobenzene 98%
1-Bromo-3-chloro-4-fluorobenzene
Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 188…194 °C
Density: 1.727 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00051794
CAS Number: 60811-21-4
EINECS: 405-580-2
UN: 3082
ADR: 9+EHS III