1-Bromo-4-chloro-2-fluorobenzene 97%
Supplier: Apollo Scientific
Warning
PC1367R-25G PC1367R-100G
APOSPC1367R-25GEA 21.9 EUR
APOSPC1367R-25G APOSPC1367R-100G
1-Bromo-4-chloro-2-fluorobenzene 97%
1-Bromo-4-chloro-2-fluorobenzene
Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 91…92 °C/20mmHg
Density: 1.678 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00079708
CAS Number: 1996-29-8