2-Bromo-4,5-difluorobenzonitrile
Supplier: Apollo Scientific
Warning
PC1369-25G PC1369-5G
APOSPC1369-25GEA 74.1 EUR
APOSPC1369-25G APOSPC1369-5G
2-Bromo-4,5-difluorobenzonitrile
2-Bromo-4,5-difluorobenzonitrile
Formula: C₇H₂BrF₂N
MW: 218.0000 g/mol
Melting Pt: 63…64 °C
Storage Temperature: Ambient
MDL Number: MFCD09864661
CAS Number: 64695-82-5