4-Bromo-3,5-dichlorobenzotrifluoride 97%
Supplier: Apollo Scientific
Warning
PC1374N-100G PC1374N-25G PC1374N-5G
APOSPC1374N-100GEA 163 EUR
APOSPC1374N-100G APOSPC1374N-25G APOSPC1374N-5G
4-Bromo-3,5-dichlorobenzotrifluoride 97%
4-Bromo-3,5-dichlorobenzotrifluoride
Formula: C₇H₂BrCl₂F₃
MW: 293.9 g/mol
Boiling Pt: 91…92 °C/10mmHg
Density: 1.833 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00221020
CAS Number: 118754-53-3