2-Bromo-4,6-difluoroaniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: BDFA
PC1376-25G PC1376-100G
APOSPC1376-25GEA 21.9 EUR
APOSPC1376-25G APOSPC1376-100G
2-Bromo-4,6-difluoroaniline 98%
2-Bromo-4,6-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Melting Pt: 38…42 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00009639
CAS Number: 444-14-4
EINECS: 429-430-0
UN: 3077
ADR: 9+EHS,III