4-Bromo-2,5-difluoroanisole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2,5-difluoro-4-methoxybenzene
PC1380D-25G PC1380D-5G
APOSPC1380D-25GEA 63.8 EUR
APOSPC1380D-25G APOSPC1380D-5G
4-Bromo-2,5-difluoroanisole 99%
4-Bromo-2,5-difluoroanisole
Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00143258
CAS Number: 202865-60-9
EINECS: 606-491-3