4-Bromo-3,5-difluoroanisole 98%
Supplier: Apollo Scientific
Warning
PC1380G-5G PC1380G-25G PC1380G-100G
APOSPC1380G-5GEA 72.5 EUR
APOSPC1380G-5G APOSPC1380G-25G APOSPC1380G-100G
4-Bromo-3,5-difluoroanisole 98%
4-Bromo-3,5-difluoroanisole
Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Melting Pt: 26…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00143268
CAS Number: 202865-61-0