2-Fluoro-3-nitrobenzoic acid 99%
Supplier: Apollo Scientific
Warning
PC1387-100G PC1387-25G PC1387-5G
APOSPC1387-100GEA 136 EUR
APOSPC1387-100G APOSPC1387-25G APOSPC1387-5G
2-Fluoro-3-nitrobenzoic acid 99%
2-Fluoro-3-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 137…139 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06797825
CAS Number: 317-46-4